5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate

Toimittaja: Apollo Scientific
Vaara

PC53047-1G PC53047-5G
APOSPC53047-1GEA 190 EUR
APOSPC53047-1G APOSPC53047-5G
5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate
5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate
Kaava: C₈H₅BrF₄O₃S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD28963743
CAS-numero: 1936290-01-5
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR