1-(Trifluoromethyl)cyclopropanamine hydrochloride 95%

Toimittaja: Apollo Scientific

PC520815-250MG PC520815-500MG
APOSPC520815-250MGEA 121 EUR
APOSPC520815-250MG APOSPC520815-500MG
1-(Trifluoromethyl)cyclopropanamine hydrochloride 95%
1-(Trifluoromethyl)cyclopropanamine hydrochloride
Kaava: C₄H₆F₃N·ClH
Storage Temperature: Jääkaappi
MDL numero: MFCD11109575
CAS-numero: 112738-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR