2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%

Toimittaja: Apollo Scientific
Vaara

PC502457-250MG PC502457-1G PC502457-5G
APOSPC502457-250MGEA 176 EUR
APOSPC502457-250MG APOSPC502457-1G APOSPC502457-5G
2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%
2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide
Kaava: C₈H₄BrF₅O
MDL numero: MFCD31561157
CAS-numero: 2244081-55-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR