Methyl amino(4-fluorophenyl)acetate hydrochloride 98%

Toimittaja: Apollo Scientific
Varoitus

PC50089-1G PC50089-5G PC50089-10G PC50089-25G
APOSPC50089-1GEA 36.6 EUR
APOSPC50089-1G APOSPC50089-5G APOSPC50089-10G APOSPC50089-25G
Methyl amino(4-fluorophenyl)acetate hydrochloride 98%
Methyl amino(4-fluorophenyl)acetate hydrochloride
Kaava: C₉H₁₁ClFNO₂
Sulamispiste: 177…179 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09152734
CAS-numero: 42718-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR