Methyl 2-(bromomethyl)-5-(trifluoromethyl)benzoate

Toimittaja: Apollo Scientific
Vaara

PC500597-1G PC500597-250MG PC500597-5G
APOSPC500597-1GEA 409 EUR
APOSPC500597-1G APOSPC500597-250MG APOSPC500597-5G
Methyl 2-(bromomethyl)-5-(trifluoromethyl)benzoate
Methyl 2-(bromomethyl)-5-(trifluoromethyl)benzoate
Kaava: C₁₀H₈BrF₃O₂
Storage Temperature: Jääkaappi
MDL numero: MFCD27931130
CAS-numero: 875895-66-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR