p-Tolyl trifluoromethanesulphonate 97%

Toimittaja: Apollo Scientific
Vaara

PC500030-5G PC500030-25G
APOSPC500030-5GEA 76.4 EUR
APOSPC500030-5G APOSPC500030-25G
p-Tolyl trifluoromethanesulphonate 97%
p-Tolyl trifluoromethanesulphonate
Kaava: C₈H₇F₃O₃S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00192507
CAS-numero: 29540-83-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR