4-Bromo-3,3,4,4-tetrafluorobutyl ethyl carbonate

Toimittaja: Apollo Scientific

PC450417-25G
APOSPC450417-25GEA 1550 EUR
APOSPC450417-25G
4-Bromo-3,3,4,4-tetrafluorobutyl ethyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl ethyl carbonate
Kaava: C₇H₉BrF₄O₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD28334470
CAS-numero: 1923065-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR