3,4,5-Trifluorifenyyliboronihappo 97%

Toimittaja: Apollo Scientific
Varoitus

PC4336-5G PC4336-100G PC4336-25G
APOSPC4336-5GEA 22 EUR
APOSPC4336-5G APOSPC4336-100G APOSPC4336-25G
3,4,5-Trifluorifenyyliboronihappo 97%
3,4,5-Trifluorifenyyliboronihappo

Very reactive catalyst to produce amides in high yieldsfrom carboxylic acids and primary or secondary aminesJ. Org. Chem., 61, 4196 (1996)

Kaava: C₆H₄BF₃O₂
Sulamispiste: 275…279 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02093069
CAS-numero: 143418-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR