3,3-Difluoropiperidin-4-ol hydrochloride 95+%

Toimittaja: Apollo Scientific

PC430135-100MG PC430135-250MG PC430135-1G
APOSPC430135-100MGEA 122 EUR
APOSPC430135-100MG APOSPC430135-250MG APOSPC430135-1G
3,3-Difluoropiperidin-4-ol hydrochloride 95+%
3,3-Difluoropiperidin-4-ol hydrochloride
Kaava: C₅H₉F₂NO·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD19686686
CAS-numero: 1334416-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR