3-Fluoro-4-phenoxyaniline hydrochloride

Toimittaja: Apollo Scientific

PC410113-1G
APOSPC410113-1GEA 169 EUR
APOSPC410113-1G
3-Fluoro-4-phenoxyaniline hydrochloride
3-Fluoro-4-phenoxyaniline hydrochloride
Kaava: C₁₂H₁₀FNO·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD25371195
CAS-numero: 1431963-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR