(3-Chloro-5-trifluoromethylpyridin-2-yloxy)acetic acid ethyl ester

Toimittaja: Apollo Scientific

PC400635-1G PC400635-250MG PC400635-500MG PC400635-2.5G PC400635-5G
APOSPC400635-1GEA 409 EUR
APOSPC400635-1G APOSPC400635-250MG APOSPC400635-500MG APOSPC400635-2.5G APOSPC400635-5G
(3-Chloro-5-trifluoromethylpyridin-2-yloxy)acetic acid ethyl ester
(3-Chloro-5-trifluoromethylpyridin-2-yloxy)acetic acid ethyl ester
Kaava: C₁₀H₉ClF₃NO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD18384847
CAS-numero: 1299607-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR