Perfluoro-15-crown-5-ether 95%

Toimittaja: Apollo Scientific
Varoitus

PC3005-1G PC3005-5G PC3005-1KG PC3005-250G PC3005-500G PC3005-750G
APOSPC3005-1GEA 80.4 EUR
APOSPC3005-1G APOSPC3005-5G APOSPC3005-1KG APOSPC3005-250G APOSPC3005-500G APOSPC3005-750G
Perfluoro-15-crown-5-ether 95%
Perfluoro-15-crown-5-ether

Potential 19F NMR imaging agent, particularly effective in brain and spinal diagnostics, USP 4,838,274. Single nmr signal; liquid at room temperature.

Kaava: C₁₀F₂₀O₅
Kiehumispiste: 146 °C
Tiheys: 1,3 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01320747
CAS-numero: 97571-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR