DFDNB (1,5-Difluori-2,4-dinitrobentseeni) 97%

Toimittaja: Apollo Scientific
Vaara

PC2770-25G PC2770-5G PC2770-100G
APOSPC2770-25GEA 121 EUR
APOSPC2770-25G APOSPC2770-5G APOSPC2770-100G
DFDNB (1,5-Difluori-2,4-dinitrobentseeni) 97%
DFDNB (1,5-Difluori-2,4-dinitrobentseeni)

Protein cross-linking agent.J. Biol. Chem., 971, 246, 6489 also: Biochem.Biophysics. Res. Commun., 1973, 53, 302.

Kaava: C₆H₂F₂N₂O₄
Sulamispiste: 72…74 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00007052
CAS-numero: 327-92-4
EINECS: 206-324-0
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR