Triphenyl(2,2,2-trifluoroethyl)phosphanium trifluoromethanesulphonate 97%

Toimittaja: Apollo Scientific
Varoitus

PC200031-1G PC200031-5G
APOSPC200031-1GEA 271 EUR
APOSPC200031-1G APOSPC200031-5G
Triphenyl(2,2,2-trifluoroethyl)phosphanium trifluoromethanesulphonate 97%
Triphenyl(2,2,2-trifluoroethyl)phosphanium trifluoromethanesulphonate
Kaava: C₂₀H₁₇F₃P·CF₃O₃S
Sulamispiste: 208…209 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD22689151
CAS-numero: 135654-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR