methyl 5-methoxypyridine-2-carboxylate 98%

Toimittaja: Apollo Scientific

OR959687-5G
APOSOR959687-5GEA 201 EUR
APOSOR959687-5G
methyl 5-methoxypyridine-2-carboxylate 98%
methyl 5-methoxypyridine-2-carboxylate
Kaava: C₈H₉NO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11975090
CAS-numero: 29681-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR