Methyl 2-ethylbenzoate 95%

Toimittaja: Apollo Scientific

OR955741-1G
APOSOR955741-1GEA 298 EUR
APOSOR955741-1G
Methyl 2-ethylbenzoate 95%
Methyl 2-ethylbenzoate
Kaava: C₁₀H₁₂O₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06203917
CAS-numero: 50604-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR