Phenylmaleic acid 95%

Toimittaja: Apollo Scientific

OR955233-1G OR955233-250MG
APOSOR955233-1GEA 354 EUR
APOSOR955233-1G APOSOR955233-250MG
Phenylmaleic acid 95%
Phenylmaleic acid
Kaava: C₁₀H₈O₄
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01075740
CAS-numero: 16110-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR