2-Pyrimidineacetic acid 98%

Toimittaja: Apollo Scientific
Varoitus

OR951464-1G
APOSOR951464-1GEA 84.7 EUR
APOSOR951464-1G
2-Pyrimidineacetic acid 98%
2-Pyrimidineacetic acid
Kaava: C₆H₆N₂O₂
Storage Temperature: Pakastin
MDL numero: MFCD04973302
CAS-numero: 66621-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR