Cycloheptylmethanamine hydrochloride 95%

Toimittaja: Apollo Scientific

OR950869-10G OR950869-1G OR950869-5G
APOSOR950869-10GEA 277 EUR
APOSOR950869-10G APOSOR950869-1G APOSOR950869-5G
Cycloheptylmethanamine hydrochloride 95%
Cycloheptylmethanamine hydrochloride
Kaava: C₈H₁₇N·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD23701552
CAS-numero: 177352-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR