Methyl 2-(propylsulphanyl)acetate 95%

Toimittaja: Apollo Scientific

OR942757-1G
APOSOR942757-1GEA 417 EUR
APOSOR942757-1G
Methyl 2-(propylsulphanyl)acetate 95%
Methyl 2-(propylsulphanyl)acetate
Kaava: C₆H₁₂O₂S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11174022
CAS-numero: 20600-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR