O-Phenylhydroxylamine hydrochloride 95%

Toimittaja: Apollo Scientific

OR940013-1G OR940013-5G
APOSOR940013-1GEA 57 EUR
APOSOR940013-1G APOSOR940013-5G
O-Phenylhydroxylamine hydrochloride 95%
O-Phenylhydroxylamine hydrochloride
Kaava: C₆H₈ClNO
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00043271
CAS-numero: 6092-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR