2,4-Dibromobutanoyl chloride, Technical Grade

Toimittaja: Apollo Scientific
Vaara

OR938772-1G OR938772-5G
APOSOR938772-1GEA 59.1 EUR
APOSOR938772-1G APOSOR938772-5G
2,4-Dibromobutanoyl chloride, Technical Grade
2,4-Dibromobutanoyl chloride
Kaava: C₄H₅Br₂ClO
Storage Temperature: Jääkaappi
MDL numero: MFCD01862994
CAS-numero: 82820-87-9
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR