H-D-Nle-OMe·HCl 98%

Toimittaja: Apollo Scientific

OR933329-10G OR933329-1G OR933329-5G
APOSOR933329-10GEA 577 EUR
APOSOR933329-10G APOSOR933329-1G APOSOR933329-5G
H-D-Nle-OMe·HCl 98%
H-D-Nle-OMe·HCl
Kaava: C₇H₁₅NO₂·HCl
Storage Temperature: Huoneenlämpö
CAS-numero: 60687-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR