Methyl 2-[(dimethylamino)methyl]benzoate 95%

Toimittaja: Apollo Scientific

OR929284-1G OR929284-250MG
APOSOR929284-1GEA 541 EUR
APOSOR929284-1G APOSOR929284-250MG
Methyl 2-[(dimethylamino)methyl]benzoate 95%
Methyl 2-[(dimethylamino)methyl]benzoate
Kaava: C₁₁H₁₅NO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13249350
CAS-numero: 27171-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR