N-Formyl-DL-phenylalanine 95%

Toimittaja: Apollo Scientific

OR924498-25G OR924498-5G
APOSOR924498-25GEA 193 EUR
APOSOR924498-25G APOSOR924498-5G
N-Formyl-DL-phenylalanine 95%
N-Formyl-DL-phenylalanine
Kaava: C₁₀H₁₁NO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00063281
CAS-numero: 4289-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR