Pyrrolidin-3-yl-ethanone hydrochloride 96%

Toimittaja: Apollo Scientific

OR921754-1G OR921754-250MG
APOSOR921754-1GEA 671 EUR
APOSOR921754-1G APOSOR921754-250MG
Pyrrolidin-3-yl-ethanone hydrochloride 96%
Pyrrolidin-3-yl-ethanone hydrochloride
Kaava: C₆H₁₁NO·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD18381078
CAS-numero: 876505-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR