(S)-tert-Butyl 3-aminoazepane-1-carboxylate

Toimittaja: Apollo Scientific
Vaara

OR918689-250MG OR918689-1G
APOSOR918689-250MGEA 109 EUR
APOSOR918689-250MG APOSOR918689-1G
(S)-tert-Butyl 3-aminoazepane-1-carboxylate
(S)-tert-Butyl 3-aminoazepane-1-carboxylate
Kaava: C₁₁H₂₂N₂O₂
MDL numero: MFCD11041390
CAS-numero: 625471-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR