4-Butylpiperidine hydrochloride 95%

Toimittaja: Apollo Scientific

OR913381-100MG OR913381-1G OR913381-250MG
APOSOR913381-100MGEA 58.4 EUR
APOSOR913381-100MG APOSOR913381-1G APOSOR913381-250MG
4-Butylpiperidine hydrochloride 95%
4-Butylpiperidine hydrochloride
Kaava: C₉H₂₀ClN
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09025421
CAS-numero: 372195-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR