Methyl 4-carbamoylbenzoate 98%

Toimittaja: Apollo Scientific

OR907970-1G OR907970-5G
APOSOR907970-1GEA 155 EUR
APOSOR907970-1G APOSOR907970-5G
Methyl 4-carbamoylbenzoate 98%
Methyl 4-carbamoylbenzoate
Kaava: C₉H₉NO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00463146
CAS-numero: 6757-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR