o-Cyanophenylethylamine hydrochloride

Toimittaja: Apollo Scientific

OR906734-1G OR906734-250MG
APOSOR906734-1GEA 1210 EUR
APOSOR906734-1G APOSOR906734-250MG
o-Cyanophenylethylamine hydrochloride
o-Cyanophenylethylamine hydrochloride
Kaava: C₉H₁₀N₂·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09878804
CAS-numero: 1159826-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR