2-Isopropylpiperidine hydrochloride 95%

Toimittaja: Apollo Scientific

OR906549-100MG OR906549-1G OR906549-250MG
APOSOR906549-100MGEA 191 EUR
APOSOR906549-100MG APOSOR906549-1G APOSOR906549-250MG
2-Isopropylpiperidine hydrochloride 95%
2-Isopropylpiperidine hydrochloride
Kaava: C₈H₁₇N·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07357628
CAS-numero: 1177339-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR