Methyl 5-amino-6-chloropyridine-3-carboxylate 98%

Toimittaja: Apollo Scientific

OR901819-1G OR901819-250MG
APOSOR901819-1GEA 99.4 EUR
APOSOR901819-1G APOSOR901819-250MG
Methyl 5-amino-6-chloropyridine-3-carboxylate 98%
Methyl 5-amino-6-chloropyridine-3-carboxylate
Kaava: C₇H₇ClN₂O₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02180841
CAS-numero: 211915-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR