3-Bromoanthranilic acid 90%

Toimittaja: Apollo Scientific
Vaara

OR61456-25G OR61456-100G OR61456-500G
APOSOR61456-25GEA 43.8 EUR
APOSOR61456-25G APOSOR61456-100G APOSOR61456-500G
3-Bromoanthranilic acid 90%
3-Bromoanthranilic acid
Kaava: C₇H₆BrNO₂
MDL numero: MFCD03618453
CAS-numero: 20776-51-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR