L-Tyrosine methyl ester hydrochloride 98%

Toimittaja: Apollo Scientific
Varoitus

OR61060-100G OR61060-500G OR61060-25G
APOSOR61060-100GEA 55.5 EUR
APOSOR61060-100G APOSOR61060-500G APOSOR61060-25G
L-Tyrosine methyl ester hydrochloride 98%
L-Tyrosine methyl ester hydrochloride
Kaava: C₁₀H₁₃NO₃
Storage Temperature: Jääkaappi
MDL numero: MFCD00012607
CAS-numero: 3417-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR