2-Bromo-1-(4-cyclopropylphenyl)ethanone

Toimittaja: Apollo Scientific
Vaara

OR55672-1G OR55672-250MG
APOSOR55672-1GEA 110 EUR
APOSOR55672-1G APOSOR55672-250MG
2-Bromo-1-(4-cyclopropylphenyl)ethanone
2-Bromo-1-(4-cyclopropylphenyl)ethanone
Kaava: C₁₁H₁₁BrO
Storage Temperature: Pakastin
MDL numero: MFCD29040962
CAS-numero: 1934754-00-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR