1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride 98%

Toimittaja: Apollo Scientific
Vaara

OR5395-100G OR5395-25G OR5395-5G
APOSOR5395-100GEA 374 EUR
APOSOR5395-100G APOSOR5395-25G APOSOR5395-5G
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride 98%
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride
Kaava: C₁₃H₁₇ClN₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012836
CAS-numero: 71617-20-4
EINECS: 275-713-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR