EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%

Toimittaja: Apollo Scientific
Vaara

OR53094-100G OR53094-25G OR53094-5G
APOSOR53094-100GEA 205 EUR
APOSOR53094-100G APOSOR53094-25G APOSOR53094-5G
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)
Kaava: C₈H₁₇N₃
Tiheys: 0,877 g/cm³ (25 °C)
Storage Temperature: Pakastin
MDL numero: MFCD00044916
CAS-numero: 1892-57-5
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR