Ethyl 3-Bromopyridine-2-carboxylate

Toimittaja: Apollo Scientific

OR470685-5G OR470685-25G
APOSOR470685-5GEA 76 EUR
APOSOR470685-5G APOSOR470685-25G
Ethyl 3-Bromopyridine-2-carboxylate
Ethyl 3-Bromopyridine-2-carboxylate
Kaava: C₈H₈BrNO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13189685
CAS-numero: 434319-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR