3-(3-Piperidyl)propyl Acetate Acetate

Toimittaja: Apollo Scientific

OR470598-5G
APOSOR470598-5GEA 1020 EUR
APOSOR470598-5G
3-(3-Piperidyl)propyl Acetate Acetate
3-(3-Piperidyl)propyl Acetate Acetate
Kaava: C₁₀H₁₉NO₂·CH₃CO₂H
Storage Temperature: Huoneenlämpö
MDL numero: MFCD23381116
CAS-numero: 1427475-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR