Cyclopentanecarboximidamide hydrochloride

Toimittaja: Apollo Scientific

OR46719-1G OR46719-250MG OR46719-500MG OR46719-2.5G
APOSOR46719-1GEA 323 EUR
APOSOR46719-1G APOSOR46719-250MG APOSOR46719-500MG APOSOR46719-2.5G
Cyclopentanecarboximidamide hydrochloride
Cyclopentanecarboximidamide hydrochloride
Kaava: C₆H₁₂N₂·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08752286
CAS-numero: 68284-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR