β-Methyllevulinic acid

Toimittaja: Apollo Scientific

OR46668-1G OR46668-250MG OR46668-500MG OR46668-5G
APOSOR46668-1GEA 220 EUR
APOSOR46668-1G APOSOR46668-250MG APOSOR46668-500MG APOSOR46668-5G
β-Methyllevulinic acid
beta-Methyllevulinic acid
Kaava: C₆H₁₀O₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00143691
CAS-numero: 6628-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR