(4R)-2,2-Dioxido-4- phenyl -1,2,3-oxathiazolidine, N-BOC protected 97%

Toimittaja: Apollo Scientific
Varoitus

OR400340-1G OR400340-250MG OR400340-5G
APOSOR400340-1GEA 271 EUR
APOSOR400340-1G APOSOR400340-250MG APOSOR400340-5G
(4R)-2,2-Dioxido-4- phenyl -1,2,3-oxathiazolidine, N-BOC protected 97%
(4R)-2,2-Dioxido-4- phenyl -1,2,3-oxathiazolidine, N-BOC protected
Kaava: C₁₃H₁₇NO₅S
Storage Temperature: Jääkaappi
MDL numero: MFCD17018793
CAS-numero: 1209467-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR