Chloroacetamidine hydrochloride

Toimittaja: Apollo Scientific
Varoitus

OR3921-25G OR3921-5G
APOSOR3921-25GEA 152 EUR
APOSOR3921-25G APOSOR3921-5G
Chloroacetamidine hydrochloride
Chloroacetamidine hydrochloride
Kaava: C₂H₆Cl₂N₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00053013
CAS-numero: 10300-69-3
EINECS: 233-675-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR