4-(Hydroksimetyyli)bentsyylialkoholi 98%

Toimittaja: Apollo Scientific

OR350022-500G
APOSOR350022-500GEA 78.9 EUR
APOSOR350022-500G
4-(Hydroksimetyyli)bentsyylialkoholi 98%
4-(Hydroksimetyyli)bentsyylialkoholi

Intermediate for organic synthesis; Synthesis of high performance resin.

Kaava: C₈H₁₀O₂
Kiehumispiste: 138…143 °C/1mmHg
Sulamispiste: 114…118 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004665
CAS-numero: 589-29-7
EINECS: 209-641-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR