2-Acetyl-3-methoxy-but-2-enoic acid methyl ester

Toimittaja: Apollo Scientific

OR346251-1G OR346251-500MG OR346251-5G
APOSOR346251-1GEA 316 EUR
APOSOR346251-1G APOSOR346251-500MG APOSOR346251-5G
2-Acetyl-3-methoxy-but-2-enoic acid methyl ester
2-Acetyl-3-methoxy-but-2-enoic acid methyl ester
Kaava: C₈H₁₂O₄
Storage Temperature: Huoneenlämpö
MDL numero: MFCD19981463
CAS-numero: 943910-00-7

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR