Ethylsulphamoyl chloride

Toimittaja: Apollo Scientific
Vaara

OR346218-1G
APOSOR346218-1GEA 0 EUR
APOSOR346218-1G
Ethylsulphamoyl chloride
Ethylsulphamoyl chloride
Kaava: C₂H₆ClNO₂S
Kiehumispiste: 86 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD12401754
CAS-numero: 16548-07-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR