2-Chloro-4-nitrophenyl isocyanate 98%

Toimittaja: Apollo Scientific
Vaara

OR33666-1G OR33666-5G
APOSOR33666-1GEA 23.4 EUR
APOSOR33666-1G APOSOR33666-5G
2-Chloro-4-nitrophenyl isocyanate 98%
2-Chloro-4-nitrophenyl isocyanate

When melted, may contain small amount of sediment.

Kaava: C₇H₃ClN₂O₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00037063
CAS-numero: 40397-95-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR