2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride 95%

Toimittaja: Apollo Scientific

OR33096-1G OR33096-5G
APOSOR33096-1GEA 338 EUR
APOSOR33096-1G APOSOR33096-5G
2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride 95%
2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride
Kaava: C₁₂H₁₂ClNO
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08056659
CAS-numero: 38061-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR