(S)-3-(2-Bromophenyl)piperidine hydrochloride

Toimittaja: Apollo Scientific

OR321521-1G OR321521-250MG
APOSOR321521-1GEA 1130 EUR
APOSOR321521-1G APOSOR321521-250MG
(S)-3-(2-Bromophenyl)piperidine hydrochloride
(S)-3-(2-Bromophenyl)piperidine hydrochloride
Kaava: C₁₁H₁₄BrN·ClH
Storage Temperature: Jääkaappi
CAS-numero: 2416257-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR