N-Benzoylethylenediamine hydrochloride 95+%

Toimittaja: Apollo Scientific

OR318004-1G OR318004-5G
APOSOR318004-1GEA 535 EUR
APOSOR318004-1G APOSOR318004-5G
N-Benzoylethylenediamine hydrochloride 95+%
Kaava: C₉H₁₃ClN₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD10511533
CAS-numero: 1502-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR